ContactUsLinkComponent
Human EGF R, His Tag (EGR-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). 1 * 100 µG
Référence ACRBEGR-H5222100UG
Fournisseur:  ACROBIOSYSTEMS
Human EGF R, His Tag (EGR-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession P00533-1). 1 * 100 µG
Référence ACRBEGR-H5222100UG
Fournisseur:  ACROBIOSYSTEMS
Frequently Bought Together