Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TRYPTIC SOY YEAST EXTRACT AGAR 1 * 500 g
Catalog # LIOF610349
Supplier:
LIOFILCHEM
Certificates
Expand image
TRYPTIC SOY YEAST EXTRACT AGAR 1 * 500 g
Catalog # LIOF610349
Supplier:
LIOFILCHEM
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Accessories for anaerobic systems, anaerobic incubation system, Anaerocult®
Supplier: Merck
VWR Chemicals, Ingrédients de base pour milieu déshydraté
Supplier: VWR Chemicals
Bioindicator, Sterikon® plus
Supplier: Merck
VWR® Dehydrated Culture Media, Powdered
Supplier: VWR Chemicals
Lames immergées pour les surfaces et les liquides, Envirocheck®
Supplier: Merck
Bouillon SSL et gélose OFS
Supplier: DOEHLER GROUP
NBB®-A, gélose
Supplier: DOEHLER GROUP
Lames de contact, VWR®
Supplier: VWR Chemicals
Accessoires
Supplier: Merck
Milieux de culture déshydratés
Supplier: Merck
VWR®, Milieux de culture, prêts à l'emploi, en plaques
Supplier: VWR Chemicals
VWR® Ready to Use Media Plates (90 mm), Irradiated
Supplier: VWR Chemicals
Système d'incubation anaérobie, accessoires pour AnaeroGen
Supplier: THERMO FISHER DIAGNOSTICS