This reference includes all the spare parts of Precellys 24 needed to be changed on a yearly basis as discribed in the user manual. 1 x white blocking plate, 2 x vaccuum joint, 1 x containment seal, 1 x antirotation tubing kit. 1 * 1 KIT
Référence 432-0246
Fournisseur: Bertin Instruments
Frequently Bought Together