Human Recombinant PDGF-BB
10 µG
N° de catalogue APOSBITP1235-10UG
Fournisseur: Apollo Scientific
Human Recombinant PDGF-BB
N° de catalogue APOSBITP1235-10UG
Fournisseur: Apollo Scientific
Numéro CAS:
Aller à
Product Details & Documents
Spécifications
Spécifications
- Cdt:10 µG
- Cat. No.:APOSBITP1235-10UG
- Synonyme de protéine:Platelet Derived Growth Factor-BB
- Species:Human
- Nom de la protéine/du peptide:PDGF-BB
- Type de protéine/peptide:Recombinant
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



