2-(4-Hydroxyphényl)ethanol 98%
Bottle, 25 g
Catalog # APOSOR2934-25G
Supplier: Apollo Scientific
2-(4-Hydroxyphényl)ethanol 98%
Catalog # APOSOR2934-25G
Supplier: Apollo Scientific
CAS Number: 501-94-0
Jump to:
Product Details & Documents
Synonyms: NSC 59876, p-HPEA, Tyrosol, 4-Hydroxyphenethyl alcohol
- CAS Number:501-94-0
- Formula:C₈H₁₀O₂
- MDL Number:MFCD00002902
- Molecular Weight:138.17 g/mol
- Boiling Pt:139 °C/1mmHg
- Melting Pt:93…95 °C
- Storage Temperature:Température ambiante
- EINECS:207-930-8
Specifications
Specifications
- Type de conditionnement:Bottle
- Cdt:25 g



