D-Cyclosérine
Bottle, 25 g
N° de catalogue APOSBIC0119-25G
Fournisseur: Apollo Scientific
Aller à
Product Details & Documents
Cycloserine interferes with the bacterial cell wall synthesis by competing with D-Alanine for incorporation into the cell wall.
Synonymes: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine
- Numéro CAS:68-41-7
- Formule:C₃H₆N₂O₂
- Numéro MDL:MFCD00005353
- Poids moléculaire:102.09 g/mol
- Point de fusion:145…147 °C (déc.)
- Température de stockage:Congélateur
- EINECS:200-688-4
Spécifications
Spécifications
- Type de conditionnement:Bottle
- Cdt:25 g
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



