Human bone marrow, unprocessed
Human unprocessed bone marrow, Fresh, Pooled, 50 mL
N° de catalogue BM008F-3
Fournisseur: Charles River Laboratories Cell Solutions
Human bone marrow, unprocessed
N° de catalogue BM008F-3
Fournisseur: Charles River Laboratories Cell Solutions
Numéro CAS:
Aller à
Product Details & Documents
Bone marrow is a major source of stem cells, which are being used in cancer research, cell studies, transplantation studies, and other research.
- Test negative for Hepatitis B and C, HIV 1/2, HTLV- I/II, HCV/HBV, WNV, Trypanasoma cruzi, and Syphilis
Bone marrow collected from healthy human volunteer donors under IRB approved informed consent, drawn from the posterior iliac crest using a syringe containing anticoagulant.
Attention: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Spécifications
Spécifications
- Format de cellule:Fresh
- Cdt:50 mL
- Donneur unique/donneurs regroupés:Pooled
- Désignation:Human unprocessed bone marrow
- Cat. No.:BM008F-3