Empty racks for tipfill system, Qualitix®
Empty racks for tipfill system, Qualitix®
Empty rack for 10/200 µl tips, to be used with tipfill system
Catalog # SOCO3070.0210ER
Supplier:   Socorex Isba
Empty racks for tipfill system, Qualitix®
Catalog # SOCO3070.0210ER
Supplier:  Socorex Isba
Empty racks for tipfill system, Qualitix®
Empty racks for tipfill system, Qualitix®
Empty rack for 10/200 µl tips, to be used with tipfill system
Price
57.30 SFr.
Log in for availability
1 * 10 ST
Product Details & Documents
These empty racks are designed to be used with Qualitix® tipfill packs.
  • Racks have smooth rounded edges and ergonomic snap latch that won’t catch gloves
  • Transparent lid enables instant view of content
  • Empty racks are re-usable and autoclavable
Specifications
  • Désignation:
    Empty rack for 10/200 µl tips, to be used with tipfill system
  • Seq:
    01