Waterproof Traps
Trap maintenance, 3048×3048 mm
Catalog # 119-0341
Supplier: JUSTRITE MANUFACTURING
Jump to:
Product Details & Documents
These waterproof traps are used to protect drums and equipment to avoid contamination or damage and also shield equipment from harsh effects of direct sunlight, sand, dust and rain.
- Brass grommets in corners to secure the tarp in place
- Lightweight vinyl coated fabric
- Use under berm products to provide additional puncture and wear-resistance
Specifications
- Désignation:Trap maintenance
- L×l:3048×3048 mm
- Seq:02