Gas purifiers
Gas purifier kit, includes FID trap and fitting size 1/8”, removes oxygen, water and organic compounds
N° de catalogue SGEA1035168
Fournisseur: Trajan Scientific and Medical
Aller à
Product Details & Documents
Gas purifiers are designed to provide fast stabilisation times to reduce gas consumption and provide clean gas to GC and GC/MS systems.
- Removal of oxygen, moisture and hydrocarbons that can impact analytical results
- Fast stabilisation times
- Improved trace analysis
- Full removal of gas impurities for different applications
Gas purifiers are an essential part of your GC analysis as contaminants in gases can significantly impact the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation. Even if carrier gas is of the highest quality, contaminants can be picked up from every part of the gas line. Therefore, a gas purifier is needed to ensure that maximum productivity is achieved.
: Connecting units and filters must be ordered separately.
: Kits include gas purifiers and mounting unit.
Documents
Trajan Scientific GC Consumables
Letteratura sul prodotto
Spécifications
- Désignation:Gas purifier kit, includes FID trap and fitting size 1/8”, removes oxygen, water and organic compounds
- Seq:08
- Cat. No.:SGEA1035168
- Fitting size:1/8"