ContactUsLinkComponent
INSTRUMAT HI-Q NANO SENSOR: FOR PARTICLES 50NM - 2UM, CONCENTRATIONS 104-109 PARTICLES/ML
Catalog # ROCERMM0002
Supplier:  ROCHE PROJECT
INSTRUMAT HI-Q NANO SENSOR: FOR PARTICLES 50NM - 2UM, CONCENTRATIONS 104-109 PARTICLES/ML
Catalog # ROCERMM0002
Supplier:  ROCHE PROJECT
Frequently Bought Together