Protease and Phosphatase Inhibitor Mini Tablets contain both protease inhibitors and phosphatase inhibitors to provide broad-spectrum complete protection from dephosphorylation and proteolytic degradation. 1 * 20 Tablet
N° de catalogue PIERA32959
Fournisseur: Thermo Fisher Scientific
Frequently Bought Together