ContactUsLinkComponent
Features • Slope front ice storage bin, for Dice Cubes, Flake, Nugget and Cubelet ice. • New sleek, contemporary styling. A perfect match to MF, MV and Prodigy Scotsman modular cube ice machines in 56cm / 22” width. • Convenient & hygienic internal i 1 * 1 items
Catalog # SCOMSB322
Supplier:  SCOTSMAN
Features • Slope front ice storage bin, for Dice Cubes, Flake, Nugget and Cubelet ice. • New sleek, contemporary styling. A perfect match to MF, MV and Prodigy Scotsman modular cube ice machines in 56cm / 22” width. • Convenient & hygienic internal i 1 * 1 items
Catalog # SCOMSB322
Supplier:  SCOTSMAN
Frequently Bought Together