ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO 69 MG/ML WATER PFI-1 CAN EFFICIENTLY BLOCK THE INTERACTION OF BET BRDS WITH H3 AND H4 ACETYLATED HISTONE TAILS. IC50 VALUE: 1.89 NM (EC50 FOR THE INHIBITION OF IL6 P 1 * 10 mg
Référence MDEXHY-16586-10MG
Fournisseur:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO 69 MG/ML WATER PFI-1 CAN EFFICIENTLY BLOCK THE INTERACTION OF BET BRDS WITH H3 AND H4 ACETYLATED HISTONE TAILS. IC50 VALUE: 1.89 NM (EC50 FOR THE INHIBITION OF IL6 P 1 * 10 mg
Référence MDEXHY-16586-10MG
Fournisseur:  MEDCHEMEXPRESS
Frequently Bought Together