ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO:25MG/MLL-778123 HYDROCHLORIDE IS AN INHIBITOR OF FPTASE AND GGPTASE-I WITH IC50 OF 2 NM AND 98 NM IN ENZYME INHIBITION DETERMINATION.IC50 VALUE: 2 NM [1]TARGET: FPTASEIN VITRO: L-778123 CAN C 1 * 10 mg
Référence MDEXHY-16273A-10MG
Fournisseur:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO:25MG/MLL-778123 HYDROCHLORIDE IS AN INHIBITOR OF FPTASE AND GGPTASE-I WITH IC50 OF 2 NM AND 98 NM IN ENZYME INHIBITION DETERMINATION.IC50 VALUE: 2 NM [1]TARGET: FPTASEIN VITRO: L-778123 CAN C 1 * 10 mg
Référence MDEXHY-16273A-10MG
Fournisseur:  MEDCHEMEXPRESS
Frequently Bought Together