ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 79 MG/ML IN DMSO (AT 25 °C), QUISINOSTAT IS AN ORALLY AVAILABLE, POTENT, HYDROXAMATE, PAN-HDACI WITH BROAD ACTIVITY IN SOLID AND HEMATOLOGIC TUMOR MODELS. QUISINOSTAT INHIB 1 * 5 mg
  MDEXHY-15433-5MG
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 79 MG/ML IN DMSO (AT 25 °C), QUISINOSTAT IS AN ORALLY AVAILABLE, POTENT, HYDROXAMATE, PAN-HDACI WITH BROAD ACTIVITY IN SOLID AND HEMATOLOGIC TUMOR MODELS. QUISINOSTAT INHIB 1 * 5 mg
  MDEXHY-15433-5MG
Frequently Bought Together