ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOAP20187 IS A SYNTHETIC, CELL-PERMEABLE LIGAND THAT CAN BE USED TO INDUCE HOMODIMERIZATION OF FUSION PROTEINS CONTAINING THE DMRB DOMAIN.TARGET: SMALL DIMERIZER DRUGSYSTEMIC AP20187 ADM 1 * 1 mg
Référence MDEXHY-13992-1MG
Fournisseur:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: 10 MM IN DMSOAP20187 IS A SYNTHETIC, CELL-PERMEABLE LIGAND THAT CAN BE USED TO INDUCE HOMODIMERIZATION OF FUSION PROTEINS CONTAINING THE DMRB DOMAIN.TARGET: SMALL DIMERIZER DRUGSYSTEMIC AP20187 ADM 1 * 1 mg
Référence MDEXHY-13992-1MG
Fournisseur:  MEDCHEMEXPRESS
Frequently Bought Together