LIQUID FORMAT IS 10MM*1ML IN WATERSOLUBILITY: H2O: 10 MG/MLEXENDIN-4(EXENATIDE), A 39 AMINO ACID PEPTIDE, ACTIVATES GLP-1 (G LUCAGON-LIKE PEPTIDE-1) RECEPTORS TO INCREASE INTRACELLULAR CAMP IN PANCREATIC ACINAR CELLS AND HAS NO EFFECT ON VIP 1 * 5 mg
Catalog # MDEXHY-13443-5MG
Supplier: MEDCHEMEXPRESS
Frequently Bought Together