ContactUsLinkComponent
SOLUBILITY: SOLUBLE IN DMSOAVASIMIBE IS A ACAT INHIBITOR INCLUING ACAT-1 AND ACAT-2IN VITRO: 1) AVASIMIBE HAS BENEFICIAL ON PLAS MA LIPIDS AND DIRECT ANTIATHEROSCLEROTIC ACTIVITY, INDEPENDENT OF LIPID EFFECTS, IN VARIOUS ANIMAL MODELS OF HYPERCHO 1 * 5 mg
Catalog # MDEXHY-13215-5MG
Supplier:  MEDCHEMEXPRESS
SOLUBILITY: SOLUBLE IN DMSOAVASIMIBE IS A ACAT INHIBITOR INCLUING ACAT-1 AND ACAT-2IN VITRO: 1) AVASIMIBE HAS BENEFICIAL ON PLAS MA LIPIDS AND DIRECT ANTIATHEROSCLEROTIC ACTIVITY, INDEPENDENT OF LIPID EFFECTS, IN VARIOUS ANIMAL MODELS OF HYPERCHO 1 * 5 mg
Catalog # MDEXHY-13215-5MG
Supplier:  MEDCHEMEXPRESS
Frequently Bought Together