ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO:39MG/MLTHZ2 IS A POTENT AND SELECTIVE CDK7 INHIBITOR WITH IN VITRO IC50 OF 13.9 NM.IC50 VALUE: 13.9 NMTARGET: CDK7 INHIBITORMORE INFORMATION CAN BE FOUND IN THE FOLLOWING PATENT: (I-18)PREPAR 1 * 10 mg
Référence MDEXHY-12280-10MG
Fournisseur:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO:39MG/MLTHZ2 IS A POTENT AND SELECTIVE CDK7 INHIBITOR WITH IN VITRO IC50 OF 13.9 NM.IC50 VALUE: 13.9 NMTARGET: CDK7 INHIBITORMORE INFORMATION CAN BE FOUND IN THE FOLLOWING PATENT: (I-18)PREPAR 1 * 10 mg
Référence MDEXHY-12280-10MG
Fournisseur:  MEDCHEMEXPRESS
Frequently Bought Together