ContactUsLinkComponent
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO ≥88MG/ML WATER AZ628 IS A NEW PAN-RAF INHIBITOR FOR BRAF, BRAFV600E, AND C-RAF-1 WITH IC50 OF 105 NM, 34 NM AND 29 NM, ALSO INHIBITS VEGFR2, DDR2, LYN, FLT1, FMS, E 1 * 25 mg
Référence MDEXHY-11004-25MG
Fournisseur:  MEDCHEMEXPRESS
LIQUID FORMAT IS 10MM*1ML IN DMSOSOLUBILITY: DMSO ≥88MG/ML WATER AZ628 IS A NEW PAN-RAF INHIBITOR FOR BRAF, BRAFV600E, AND C-RAF-1 WITH IC50 OF 105 NM, 34 NM AND 29 NM, ALSO INHIBITS VEGFR2, DDR2, LYN, FLT1, FMS, E 1 * 25 mg
Référence MDEXHY-11004-25MG
Fournisseur:  MEDCHEMEXPRESS
Frequently Bought Together