Disinfectant spray, Contec® HydroPure
Contec® HydroPure, 6% hydrogen peroxide and purified water, trigger spray, Case of 6× 1 l trigger spray bottles
  CONTFBT16HP
 :   CONTEC
Disinfectant spray, Contec® HydroPure
  CONTFBT16HP
 :  CONTEC
Disinfectant spray, Contec® HydroPure
Disinfectant spray, Contec® HydroPure
Contec® HydroPure, 6% hydrogen peroxide and purified water, trigger spray, Case of 6× 1 l trigger spray bottles
SFr. 177.00
1 * 6 ST
Contec® HydroPure a blend of 6% stabilised hydrogen peroxide and purified water and leaves little to no residue and is ideal for use on most surfaces. Contec® HydroPure is BPR authorised at EU level and is efficacious against bacteria, fungi, moulds and yeasts.
  • Hydrogen peroxide breaks down to water and oxygen making it very low residue
  • Filtered to 0,2 μm under Grade A airflow in a Grade B cleanroom ensuring the product is free from contamination and particulates
  • Tigger spray and 'bag in bottle' protected system prevents bottle contents from becoming contaminated during use
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
  • Double bagged in linear tear packaging so each bag is easy to open even when wearing gloves
  • Not classed as corrosive and can be used safely in all areas of the cleanroom
The product is 0,2 μm filtered and filled under Grade A airflow in a Grade B cleanroom environment ensures the product is free from contamination and particulates.
 : A sterile version for more critical areas is also available.
 : Contec HydroPure is available in a 1 litre trigger spray and 1 litre capped container as well as a 5 litre capped container.
 : Double bagged packed in linear tear packaging to aid the transfer of the product into a pharmaceutical cleanroom and also easy to open even when wearing gloves.
 : Utilisez les biocides en toute sécurité. Toujours lire l'étiquette et les informations du produit avant utilisation.
  • Désignation:
    Contec® HydroPure, 6% hydrogen peroxide and purified water, trigger spray
  • Seq:
    03
  • Emballage:
    Case of 6× 1 l trigger spray bottles