Low noise data acquisition system plus HumSilencer, Digidata®
Low noise data acquisition system with one HumSilencer channel
N° de catalogue MLDVDIGIDATA1550B1
Fournisseur: Molecular Devices
Low noise data acquisition system plus HumSilencer, Digidata®
N° de catalogue MLDVDIGIDATA1550B1
Fournisseur: Molecular Devices
Aller à
Product Details & Documents
The low noise data acquisition system plus HumSilencer™ adaptive noise cancellation with the added benefit of single-click elimination of 50/60 Hz line-frequency noise up to four channels.
- Built-in, softwarecontrolled technology
- Removes local line-frequency noise patterns
- Associated high-frequency harmonics from incoming signals in less than one second
Intended for precision scientific applications, it is particularly designed for electrophysiology experiments, to send and receive signals from microelectrode amplifiers, and to interact with peripheral instruments such as solution exchangers.
Documents
TDS Axon Digidata 1550B plus HumSilencer
Fiche technique
Spécifications
- Désignation:Low noise data acquisition system with one HumSilencer channel
- Seq:02