Low noise data acquisition system plus HumSilencer, Digidata®
Low noise data acquisition system plus HumSilencer, Digidata®
Low noise data acquisition system without HumSilencer
Catalog # MLDVDIGIDATA1550B0
Supplier:   Molecular Devices
Low noise data acquisition system plus HumSilencer, Digidata®
Catalog # MLDVDIGIDATA1550B0
Supplier:  Molecular Devices
Low noise data acquisition system plus HumSilencer, Digidata®
Low noise data acquisition system plus HumSilencer, Digidata®
Low noise data acquisition system without HumSilencer
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The low noise data acquisition system plus HumSilencer™ adaptive noise cancellation with the added benefit of single-click elimination of 50/60 Hz line-frequency noise up to four channels.
  • Built-in, softwarecontrolled technology
  • Removes local line-frequency noise patterns
  • Associated high-frequency harmonics from incoming signals in less than one second
Intended for precision scientific applications, it is particularly designed for electrophysiology experiments, to send and receive signals from microelectrode amplifiers, and to interact with peripheral instruments such as solution exchangers.
Specifications
  • Désignation:
    Low noise data acquisition system without HumSilencer
  • Seq:
    01