Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Light boxes, Slimlite plano LED

Light boxes, Slimlite plano LED

Light box, illumination area 320×228 mm, 4,1 W, W×D×H 390×270×8 mm, 1010 g
N° de catalogue KAFO2454
Fournisseur:   KAISER FOTOTECHNIK
Light boxes, Slimlite plano LED
N° de catalogue KAFO2454
Fournisseur:  KAISER FOTOTECHNIK
Light boxes, Slimlite plano LED
Light boxes, Slimlite plano LED

Light box, illumination area 320×228 mm, 4,1 W, W×D×H 390×270×8 mm, 1010 g

Prix
SFr. 284.00
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Super flat and dimmable daylight light box with build-in rechargeable battery.

  • Uniform and colour precise illumination for viewing, sorting or digitising slides or other transparent originals
  • Various power supply options: Built-in lithium ion battery, the combined mains adapter/charger included or USB cable using a powerbank, computer or car
  • Colour temperature ~5000 Kelvin and colour renditon index of 88 provides for precise colour rendition for inspection and photographic reproduction of originals
  • Scale in cm and inches beside the illuminated area
  • A short touch of the touch switch activates the light source, a longer touch changes brightness continuously

Light source: 23 LEDs, 5000 Kelvin, CRI=88
Built-in Lithium-ion battery (1050 mA) has an operating time of ~6 hours at full power, longer when dimmed.

Spécifications
  • Désignation:
    Light box, illumination area 320×228 mm, 4,1 W, W×D×H 390×270×8 mm, 1010 g
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..