Spray chambers and brackets
Replacement helix locking screw for ICP spray chamber, suits any ICP spray chamber that uses helix fitting to secure nebulizer in spray chamber side arm
N° de catalogue HEWLG8010-80043
Fournisseur: AGILENT HEWLETT PACKARD
Aller à
Product Details & Documents
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Spécifications
- Désignation:Replacement helix locking screw for ICP spray chamber, suits any ICP spray chamber that uses helix fitting to secure nebulizer in spray chamber side arm
- Seq:09