MP-AES torches | Avantor
MP-AES torches
Ball joint adaptor, PTFE, for Agilent 4200 MP-AES inert torch, required to connect alumina injector of the inert torch to the inert spray chamber
N° de catalogue HEWLG8003-20011
Fournisseur:   AGILENT HEWLETT PACKARD
MP-AES torches
N° de catalogue HEWLG8003-20011
MP-AES torches
MP-AES torches
Ball joint adaptor, PTFE, for Agilent 4200 MP-AES inert torch, required to connect alumina injector of the inert torch to the inert spray chamber
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
MP-AES Torches for the 4100/4200 Microwave Plasma-Atomic Emission Spectrometer.
  • Accuracy even with difficult samples — the vertical plasma torch provides excellent performance for tough samples with axial viewing for the best sensitivity
  • Reduce sample preparation
  • The axially viewed vertical plasma handles your toughest samples — from food and soil digests to soil extractions
  • Improve productivity when running hydrofluoric (HF) acid digests
  • The inert torch enables direct analysis of HF acid digests directly without a prior neutralization step
The Agilent 4200 MP-AES features a plug and play torch, optimized for high precision and sensitivity with the recommended power and gas flow settings. The torch features a unique polymer base that fits easily into the simple and effective torch-loader mechanism. This automatically aligns the torch and connects the gases to ensure fast start-up and reproducible performance. Once loaded, there is no need for further adjustments of the torch. Automatic alignment ensures reproducible performance is achieved with each operator, and helps to greatly reduce instrument to instrument variability.Agilent’s 4200 MP-AES also features a robust vertical torch with end-on axial viewing for excellent detection limits. This means uncompromised measurements and best performance with difficult samples such as those with high total dissolved solids. The vertical orientation also ensures reduced cleaning and downtime.
Spécifications
  • Désignation:
    Ball joint adaptor, PTFE, for Agilent 4200 MP-AES inert torch, required to connect alumina injector of the inert torch to the inert spray chamber
  • Seq:
    01