BUFFER SOLUTION PH1.68 / 0.01, 25C 1 * 500 mL
BUFFER SOLUTION PH1.68 / 0.01, 25C 1 * 500 mL
Catalog # REGEEP168
Supplier:   REAGECON
BUFFER SOLUTION PH1.68 / 0.01, 25C 1 * 500 mL
Catalog # REGEEP168
Supplier:  REAGECON
BUFFER SOLUTION PH1.68 / 0.01, 25C 1 * 500 mL
Price
SFr. 69.10
Log in for availability
1 * 500 mL
Product Details & Documents