Drum racks
Drum racks
Type 400, with GRP sump pallet (220 l), barrel support for 2×60 l or 1×200 l barrels and grating support for small containers, heights of storage levels from ground 620/1390 mm, 82 kg, 990×1280×1620 mm
Catalog # CEMG7665
Supplier:   CEMO
Drum racks
Catalog # CEMG7665
Supplier:  CEMO
Drum racks
Drum racks
Type 400, with GRP sump pallet (220 l), barrel support for 2×60 l or 1×200 l barrels and grating support for small containers, heights of storage levels from ground 620/1390 mm, 82 kg, 990×1280×1620 mm
Price
SFr. 1’230.00
Log in for availability
1 * 1 ST
Product Details & Documents
Drum racks for approved and space saving storage of barrels and other containers.
  • All drum racks are completely hotdip galvanised and have a corrosion free GRP sump pallet for long-term safe use
  • With general construction inspection approval no. Z-40.12-227 granted by DIBt-Berlin
  • Universally applicable for waste oil, lyes and acids
Specifications
  • Poids:
    82 kg
  • Désignation:
    Type 400, with GRP sump pallet (220 l), barrel support for 2×60 l or 1×200 l barrels and grating support for small containers, heights of storage levels from ground 620/1390 mm
  • Seq:
    07
  • l×p×h:
    990×1280×1620 mm
Accessories
Product Information
  • Catalog #
    Désignation
    Poids
    Availability
    Price
  • CEMG5917
    Can holder, galvanised, for drum racks type 120/360/540, with surface 230×110 mm
    2 kg
    Log in for availability
    1 * 1 ST - SFr. 49.20
Product Information
  • Catalog #
    Désignation
    Poids
    Availability
    Price
  • CEMG7669
    Rotating roller support for drum racks type 400/800 (for 200 l barrel (pair), heights of storage levels 660 mm from ground
    6 kg
    Log in for availability
    1 * 1 ST - SFr. 282.00
Product Information
  • Catalog #
    Désignation
    Poids
    Availability
    Price
  • CEMG8254
    Drum supports for drum racks for type 360/540, 3 pieces for one storage level for optimum emptying of the 60 l drums
    1 kg
    Log in for availability
    1 * 3 ST - SFr. 61.60
Product Information
  • Catalog #
    Désignation
    Poids
    Availability
    Price
  • CEMG6817
    Steel grating for drum racks, as 3rd (for type 360) or 4th storage level (for type 540), 1300×620×30 mm, 210 mm storage height from ground
    18 kg
    Log in for availability
    1 * 1 ST - SFr. 153.00
Product Information
  • Catalog #
    Désignation
    Poids
    Availability
    Price
  • CEMG7727
    Can holder, galvanised, for drum racks type 400/800, with bearing surface 185×110 mm
    2 kg
    Log in for availability
    1 * 1 ST - SFr. 56.20
Product Information
  • Catalog #
    Désignation
    Poids
    Availability
    Price
  • CEMG7800
    Drum supports, 3 pieces per storage level, for drum racks type 360/540
    1 kg
    Log in for availability
    1 * 1 ST - SFr. 57.90
Product Information
  • Catalog #
    Désignation
    Poids
    l×p×h
    Availability
    Price
  • CEMG7871
    Sump pallet, PE, 40 l, without grating
    5 kg
    1300×620×80 mm
    Log in for availability
    1 * 1 ST - SFr. 196.00