Thread adapter, GL PTFE , BOLA
With 2 GL threads, GL18 (from male thread) and GL25 (to male thread), 10,5 mm
N° de catalogue BOHLD537-12
Fournisseur: Bohlender
Aller à
Product Details & Documents
Straight transition thread adapter made of PTFE with two GL-threaded connections.
- Chemical resistant
- Vacuum suitable
- Temperature resistant from –200 to +250 °C
Spécifications
- Ø trou:10,5 mm
- Désignation:With 2 GL threads, GL18 (from male thread) and GL25 (to male thread)