Extraction Buffer 5X PTR, Abcam
Extraction Buffer 5X PTR, Abcam
Extraction Buffer 5X PTR
N° de catalogue ABCAAB193970-50
Marketsource
Fournisseur:   Abcam
Extraction Buffer 5X PTR, Abcam
N° de catalogue ABCAAB193970-50
Fournisseur:  Abcam
Marketsource
Numéro CAS:  
Extraction Buffer 5X PTR, Abcam
Extraction Buffer 5X PTR
Numéro CAS:  
Prix
69.90 SFr.
Connexion Pour la disponibilité
1 * 50 mL
Product Details & Documents
Extraction buffer 5X PTR is formulated for preparing cell or tissue extracts for use in immunoassays, with added phosphatase inhibitors and protease inhibitor aprotinin. - Cited in over 20 publications - Phosphatase and protease inhibitors – contains inhibitors to preserve sample integrity - Easy to use – dilute to 1X with distilled water before use, or use with extraction enhancer buffer 50X for enhanced extraction - Compatible with a variety of sample types
Attention: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Spécifications
Spécifications
  • Désignation:
    Extraction Buffer 5X PTR
  • Cat. No.:
    ABCAAB193970-50
  • Température de stockage:
    Store at +4°C.