Sterilisation Boxes for Pipettes
Pipette box for the storage of pipettes (150 - 200 mm)
N° de catalogue BRND28920
Fournisseur: Brand
Aller à
Product Details & Documents
These pipette boxes made of aluminum are ideal for the storage and sterilisation of pipettes with different lengths.
- With inner silicone coating
- Very good temperature resistance
- Very good chemical resistance
Spécifications
- Désignation:Pipette box for the storage of pipettes (150 - 200 mm)
- Seq:01