4-Hydroxybenzoate d'éthyle NF
Poly pail, 12 kg
Catalog # SPCMET117-12KG
Supplier: Spectrum Chemical
4-Hydroxybenzoate d'éthyle NF
Catalog # SPCMET117-12KG
Supplier: Spectrum Chemical
CAS Number: 120-47-8
Jump to:
Product Details & Documents
Synonyms: Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
- CAS Number:120-47-8
- Formula:C₉H₁₀O₃
- MDL Number:MFCD00002353
- Molecular Weight:166.18 g/mol
- Merck Index:13,03869
- Boiling Pt:297 °C (1013 hPa)
- Melting Pt:116…118 °C
- Density:1,078 g/cm³ (131 °C)
- Storage Temperature:Température ambiante
- EINECS:204-399-4
Specifications
Specification Test Results
- Assay:98% Min.
Specifications
- Type de conditionnement:Poly pail
- Cdt:12 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



