SONOREX TECHNIK RM 75 UH/40 kHz, cuve ultrasonique, dim.inter. 580x500x300 mm 90,0 l 230 V( 10%) 50/60 Hz, minuterie 1-15 et en continue chauffage 30-80C, cuve soude 2mm, 1.4571, puissance 1000 W eff. 1 * 1 Pce
Catalog # BAND8220-GB
Anbieter: BANDELIN-ELECTRONIC
Frequently Bought Together