Biopsy processing/embedding cassettes, Unisette™ M406
turquoise
N° de catalogue SIMPM406-12T
Fournisseur: Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
N° de catalogue SIMPM406-12T
Fournisseur: Simport Scientific
Aller à
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
- Moulded from a special high density polymer
- Resistant to the chemical action of most histological solvents
Documents
SIMPM406-7_TDS_EN
Fiche technique
Spécifications
- Couleur:turquoise
- Seq:21