Biopsy processing/embedding cassettes, Unisette™ M406
orange
N° de catalogue SIMPM406-11
Fournisseur: Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
N° de catalogue SIMPM406-11
Fournisseur: Simport Scientific
Aller à
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
- Moulded from a special high density polymer
- Resistant to the chemical action of most histological solvents
Documents
Spécifications
- Couleur:orange
- Seq:10