Biopsy processing/embedding cassettes, Unisette™ M406
jaune
Katalognr. SIMPM406-5
Anbieter: Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Katalognr. SIMPM406-5
Anbieter: Simport Scientific
Direkt zu
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
- Moulded from a special high density polymer
- Resistant to the chemical action of most histological solvents
Dokumente
SIMPM406-7_TDS_EN
Technisches Datenblatt
Technische Daten
- Couleur:jaune
- Seq:04