MultiScreen® HTS Filter Plates, Styrene
Multiscreen HTS filter plate, Polystyrène, 50 - 250 μl, 96, -
N° de catalogue MILFMSNANLY50
Fournisseur: Merck
Aller à
Product Details & Documents
MultiScreen plates have a long history as a reliable tool for the advancement of drug discovery and life science research.
- Enhanced handling is achieved with rigid sidewalls
- Abundant surface area for barcode labels
- Prevents any cross-contamination during incubation
- High throughput screening
- Color code: Clear
MultiScreen HTS plates are specifically developed for high throughput use with automated workstations. Plasmid purification is a widely employed method in molecular cloning, transformation, and recombinant protein expression. Plasmid purification kits are designed to accommodate different bacterial culture sizes and the corresponding plasmid yield.
Multiscreen HTS filter plate, styrene is suitable for use in the clearing of bacterial lysates with the MultiScreen HTS vacuum manifold prior to plasmid purification using MILFMSNUPSD50.
Spécifications
- Nb de puits:96
- Stérile:-
- Désignation:Multiscreen HTS filter plate
- Matière:Polystyrène
- Seq:02
- Volume puits:50 - 250 μl