Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
Film applicators, Baker

Film applicators, Baker

Baker film applicator, 4-sided, gaps 90, 90, 150, 150 µm, width 80 mm
Catalog # SIMEVF1560
Supplier:   Industrial Physics
Film applicators, Baker
Catalog # SIMEVF1560
Supplier:  Industrial Physics
Film applicators, Baker
Film applicators, Baker

Baker film applicator, 4-sided, gaps 90, 90, 150, 150 µm, width 80 mm

Price
SFr. 844.00
Log in for availability
1 * 1 ST
Product Details & Documents

Cylindrical film applicator with four application sides for applying paint films of four different predefined thicknesses in film width 60 or 80 mm. Suitable for applying a host of different products onto flat and relatively solid substrates. Made from high grade stainless steel, the film applicator will not be affected by acid or base elements.

Delivery information: Supplied in a plastic case with calibration certificate.

Documents

Specifications
  • Désignation:
    Baker film applicator, 4-sided, gaps 90, 90, 150, 150 µm, width 80 mm
  • Seq:
    17
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.