Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Film applicators, Baker

Baker film applicator, 4-sided, gaps 30, 60, 90, 120 µm, width 100 mm
Catalog # SIMEVF1522
Supplier:   Industrial Physics
Film applicators, Baker
Catalog # SIMEVF1522
Supplier:  Industrial Physics
Film applicators, Baker
Film applicators, Baker

Baker film applicator, 4-sided, gaps 30, 60, 90, 120 µm, width 100 mm

Price
SFr. 899.00
Log in for availability
1 * 1 ST
Product Details & Documents

Cylindrical film applicator with four application sides for applying paint films of four different predefined thicknesses in film width 60 or 80 mm. Suitable for applying a host of different products onto flat and relatively solid substrates. Made from high grade stainless steel, the film applicator will not be affected by acid or base elements.

 : Supplied in a plastic case with calibration certificate.

Specifications
  • Désignation:
    Baker film applicator, 4-sided, gaps 30, 60, 90, 120 µm, width 100 mm
  • Seq:
    11