Film applicators, Baker
Baker film applicator, 4-sided, gaps as desired, width 100 mm
SIMEVF1520
Cylindrical film applicator with four application sides for applying paint films of four different predefined thicknesses in film width 60 or 80 mm. Suitable for applying a host of different products onto flat and relatively solid substrates. Made from high grade stainless steel, the film applicator will not be affected by acid or base elements.
: Supplied in a plastic case with calibration certificate.
- Désignation:Baker film applicator, 4-sided, gaps as desired, width 100 mm
- Seq:09