Film applicators, Baker
Baker film applicator, 4-sided, gaps as desired, width 100 mm
  SIMEVF1520
Film applicators, Baker
  SIMEVF1520
Film applicators, Baker
Film applicators, Baker
Baker film applicator, 4-sided, gaps as desired, width 100 mm
1 010.00 SFr.
1 * 1 ST
Cylindrical film applicator with four application sides for applying paint films of four different predefined thicknesses in film width 60 or 80 mm. Suitable for applying a host of different products onto flat and relatively solid substrates. Made from high grade stainless steel, the film applicator will not be affected by acid or base elements.
 : Supplied in a plastic case with calibration certificate.
  • Désignation:
    Baker film applicator, 4-sided, gaps as desired, width 100 mm
  • Seq:
    09