Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Tigecycline, Reference Standards

Tigecycline, USP reference standard, Ampule, 300 mg
N° de catalogue 1667643.
Fournisseur:   USP
Tigecycline, Reference Standards
N° de catalogue 1667643.
Fournisseur:  USP
Numéro CAS:  
Tigecycline, Reference Standards

Tigecycline, USP reference standard, Ampule, 300 mg

Numéro CAS:  
Prix
SFr. 823.00
Connexion Pour la disponibilité
1 * 300 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

 : Cold Shipment required

Spécifications

Spécifications

  • Type de conditionnement:
    Ampule
  • Cdt:
    300 mg
  • Normes:
    USP reference standard
  • Désignation:
    Tigecycline
  • UN No.:
    3077
  • Cat. No.:
    1667643.
  • N ° CAS.:
    220620-09-7
  • Formula:
    C₂₉H₃₉N₅O₈
  • Désignation:
    Tigecycline

Spécifications