Tubes and systems for active sampling
Activated charcoal tubes, BIA
N° de catalogue DRAE6400743
Fournisseur: Dräger
Aller à
Product Details & Documents
Sampling activated tubes for direct measurement indication even with complex compounds and mixtures of substances in air investigations and gas analyses.
- High precision guaranteed
- Very reliable in use
- Improved colour depth and contrast using an optimised production method
Spécifications
- Type:BIA
- Désignation:Activated charcoal tubes
- Seq:03