Potassium hydroxyde 1 N dans l'eau
Flacon plastique, 2,5 L
: Potash caustic, Caustic potash
- :1310-58-3
- :KOH
- :MFCD00003553
- :56.11 g/mol
- :100 °C (760 mmHg)
- :1,05 g/cm³
- :Température ambiante
- :1814
- :8,II
- Appearance:Clear colorless Solution
- Strength:1 N Exact strength on the certificate of analysis
- Type de conditionnement:Flacon plastique
- Cdt:2,5 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



