Résorcinol 98%
Plastic drum, 5 kg
: 1,3-Dihydroxybenzene, 1,3-Benzenediol
- :108-46-3
- :C₆H₆O₂
- :MFCD00002269
- :110.11 g/mol
- :12,08323
- :281 °C (760 mmHg)
- :109…111 °C
- :1,272 g/cm³ (20 °C)
- :127 °C
- :Température ambiante
- :203-585-2
- :2876
- :6.1,III
- Appearance:White to yellow to beige to pink Crystals or granules or flakes
- Infrared spectrum:Conforms
- Melting point:108 to 112 °C
- GC:≥97.5 %
- Type de conditionnement:Plastic drum
- Cdt:5 kg



