Saisie de commande
Switzerland
Lien pour commandesContactUsLinkComponent
 

Résorcinol 98%

Flacon plastique, 1 kg
Catalog # ACRO132290010
Irritant
Environment
Résorcinol 98%
Catalog # ACRO132290010
CAS Number:  108-46-3
Résorcinol 98%
Résorcinol 98%

Flacon plastique, 1 kg

CAS Number:  108-46-3
Price
SFr. 144.00
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  1,3-Dihydroxybenzene, 1,3-Benzenediol
  • CAS Number:108-46-3
  • Formula:C₆H₆O₂
  • MDL Number:MFCD00002269
  • Molecular Weight:110.11 g/mol
  • Merck Index:12,08323
  • Boiling Pt:281 °C (760 mmHg)
  • Melting Pt:109…111 °C
  • Density:1,272 g/cm³ (20 °C)
  • Flash Pt:127 °C
  • Storage Temperature:Température ambiante
  • EINECS:203-585-2
  • UN:2876
  • ADR:6.1,III
Specifications

Specification Test Results

  • Appearance:
    White to yellow to beige to pink Crystals or granules or flakes
  • Infrared spectrum:
    Conforms
  • Melting point:
    108 to 112 °C
  • GC:
    ≥97.5 %

Specifications

  • Type de conditionnement:
    Flacon plastique
  • Cdt:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.