Fluorescéine, pur
Flacon Verre, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 2-(6-Hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid
- CAS Number:2321-07-5
- Formula:C₂₀H₁₂O₅
- MDL Number:MFCD00005050
- Molecular Weight:332.31 g/mol
- Merck Index:13,04185
- Melting Pt:314…316 °C
- Storage Temperature:Température ambiante
- EINECS:219-031-8
Documents
Specifications
Specification Test Results
- Appearance:Red-brown to rust brown to brown Powder
- Infrared spectrum:Conforms
- Loss on drying:≤3 %
- Sulfated ash:≤0.5 %
- Specific absorbance A (1%/1cm):≥1800 (490 +/- 5 nm , 0.5% aq. soln. of Na2CO3)
Specifications
- Type de conditionnement:Flacon Verre
- Cdt:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



