Squeegee mop and handles
Squeegee mop, white, 560 mm
N° de catalogue PPSA3400715
Fournisseur: PFENNIG REINIGUNGSTECHNIK
Aller à
Product Details & Documents
Squeegee mops and handles for hygienically sensitive areas.
- Mop: PP (body), TPE (lip)
- Hygienic squeegee is suitable with cleanroom and telescopical handles
- Handles are anodised aluminum tubes
Spécifications
- Désignation:Squeegee mop, white, 560 mm
- Seq:01