Inserts
E 907/1, insert, mesh tray with lid, for small items, mesh size: 3×1 mm, PG 8535/36, 129×170×48 mm
N° de catalogue MIEL6943820
Fournisseur: MIELE
Aller à
Product Details & Documents
Spécifications
- Désignation:E 907/1, insert, mesh tray with lid, for small items, mesh size: 3×1 mm
- Seq:21
- l×p×h:129×170×48 mm
- Pour:PG 8535/36
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



