ContactUsLinkComponent
Converts Liquid waste to solid waste for easy disposal.  Super absorbent granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste.  Common uses: Put Lab-Sorb at the bottom of biohazard bin, Liquid containment for spills, convert 1 * 1 Pce
Référence DIVESORB-1000
Fournisseur:  DIVERSIFIED BIOTECH
Converts Liquid waste to solid waste for easy disposal.  Super absorbent granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste.  Common uses: Put Lab-Sorb at the bottom of biohazard bin, Liquid containment for spills, convert 1 * 1 Pce
Référence DIVESORB-1000
Fournisseur:  DIVERSIFIED BIOTECH
Frequently Bought Together